2-Bromo-4-fluoro-6-iodoaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC501968-1G
PC501968-250MG
APOSPC501968-1GEA
12400
CZK
APOSPC501968-1G
APOSPC501968-250MG
2-Bromo-4-fluoro-6-iodoaniline
2-Bromo-4-fluoro-6-iodoaniline
Vzorec:
C₆H₄BrFIN Storage Temperature: Ambient |
MDL Number:
MFCD24160442 CAS číslo: 1447606-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...