Tisk…

5-(Difluoromethoxy)-2-fluoronitrobenzene 95%

Dodavatel: Apollo Scientific

PC501967-1G PC501967-250MG PC501967-5G
APOSPC501967-1GEA 16370 CZK
APOSPC501967-1G APOSPC501967-250MG APOSPC501967-5G
5-(Difluoromethoxy)-2-fluoronitrobenzene 95%
5-(Difluoromethoxy)-2-fluoronitrobenzene
Vzorec: C₇H₄F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD14698454
CAS číslo: 57213-30-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR