Tisk…

3-Amino-4-(trifluoromethoxy)benzoic acid

Dodavatel: Apollo Scientific
Varování

PC501957-1G PC501957-250MG
APOSPC501957-1GEA 1990 CZK
APOSPC501957-1G APOSPC501957-250MG
3-Amino-4-(trifluoromethoxy)benzoic acid
3-Amino-4-(trifluoromethoxy)benzoic acid
Vzorec: C₈H₆F₃NO₃
MW: 221,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390052
CAS číslo: 656-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR