2-Bromo-3,6-difluorotoluene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-1,4-difluoro-3-methylbenzene
PC501951-1G
PC501951-250MG
PC501951-5G
APOSPC501951-1GEA
4960
CZK
APOSPC501951-1G
APOSPC501951-250MG
APOSPC501951-5G
2-Bromo-3,6-difluorotoluene
2-Bromo-3,6-difluorotoluene
Vzorec:
C₇H₅BrF₂ MW: 207,015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12922624 CAS číslo: 1208074-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...