Tisk…

3-Bromo-5-fluoro-2-nitroaniline 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC501917-1G PC501917-250MG PC501917-5G
APOSPC501917-1GEA 6620 CZK
APOSPC501917-1G APOSPC501917-250MG APOSPC501917-5G
3-Bromo-5-fluoro-2-nitroaniline 95%
3-Bromo-5-fluoro-2-nitroaniline
Vzorec: C₆H₄BrFN₂O₂
MW: 235,011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739824
CAS číslo: 1350117-42-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR