Tisk…

2-Bromo-3,5-difluorobenzyl bromide 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC501911-1G PC501911-250MG PC501911-5G
APOSPC501911-1GEA 953 CZK
APOSPC501911-1G APOSPC501911-250MG APOSPC501911-5G
2-Bromo-3,5-difluorobenzyl bromide 95%
2-Bromo-3,5-difluorobenzyl bromide
Vzorec: C₇H₄Br₂F₂
Storage Temperature: Ambient
MDL Number: MFCD28737918
CAS číslo: 1807071-26-6
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR