2-(Difluoromethoxy)-6-fluoroaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC501904-1G
PC501904-250MG
PC501904-5G
APOSPC501904-1GEA
11740
CZK
APOSPC501904-1G
APOSPC501904-250MG
APOSPC501904-5G
2-(Difluoromethoxy)-6-fluoroaniline
2-(Difluoromethoxy)-6-fluoroaniline
Vzorec:
C₇H₆F₃NO Storage Temperature: Refrigerator |
MDL Number:
MFCD18385074 CAS číslo: 1261839-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...