Tisk…

4-(Difluoromethoxy)-3-fluoroaniline

Dodavatel: Apollo Scientific
Varování

PC501893-1G PC501893-250MG PC501893-5G
APOSPC501893-1GEA 705 CZK
APOSPC501893-1G APOSPC501893-250MG APOSPC501893-5G
4-(Difluoromethoxy)-3-fluoroaniline
4-(Difluoromethoxy)-3-fluoroaniline
Vzorec: C₇H₆F₃NO
MW: 177,124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11621754
CAS číslo: 83190-01-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR