2-Amino-3-bromo-5-fluorophenol
Dodavatel: Apollo Scientific
|
Varování
|
PC501881-1G
PC501881-5G
APOSPC501881-1GEA
11580
CZK
APOSPC501881-1G
APOSPC501881-5G
2-Amino-3-bromo-5-fluorophenol
2-Amino-3-bromo-5-fluorophenol
Vzorec:
C₆H₅BrFNO MW: 206,012 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28738823 CAS číslo: 1805027-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...