Tisk…

2-Bromo-3,5-difluorobenzoic acid

Dodavatel: Apollo Scientific
Varování

PC501854-1G PC501854-5G PC501854-25G
APOSPC501854-1GEA 622 CZK
APOSPC501854-1G APOSPC501854-5G APOSPC501854-25G
2-Bromo-3,5-difluorobenzoic acid
2-Bromo-3,5-difluorobenzoic acid
Vzorec: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835211
CAS číslo: 651027-01-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR