Tisk…

4-Bromo-2,5-difluorobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

PC501849-1G PC501849-250MG PC501849-5G
APOSPC501849-1GEA 20830 CZK
APOSPC501849-1G APOSPC501849-250MG APOSPC501849-5G
4-Bromo-2,5-difluorobenzyl bromide
4-Bromo-2,5-difluorobenzyl bromide
Vzorec: C₇H₄Br₂F₂
MW: 285,911 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847141
CAS číslo: 486460-10-0
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR