2-Bromo-3-chloro-6-fluorobenzonitrile
Dodavatel: Apollo Scientific
|
Varování
|
PC501834-1G
PC501834-250MG
APOSPC501834-1GEA
26880
CZK
APOSPC501834-1G
APOSPC501834-250MG
2-Bromo-3-chloro-6-fluorobenzonitrile
2-Bromo-3-chloro-6-fluorobenzonitrile
Vzorec:
C₇H₂BrClFN Storage Temperature: Ambient |
MDL Number:
MFCD28141935 CAS číslo: 1823576-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...