Tisk…

5-Bromo-3-nitro-2-(trifluoromethyl)pyridine 95%

Dodavatel: Apollo Scientific

PC50183-1G PC50183-250MG PC50183-5G
APOSPC50183-1GEA 14850 CZK
APOSPC50183-1G APOSPC50183-250MG APOSPC50183-5G
5-Bromo-3-nitro-2-(trifluoromethyl)pyridine 95%
5-Bromo-3-nitro-2-(trifluoromethyl)pyridine
Vzorec: C₆H₂BrF₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18255263
CAS číslo: 1211540-75-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR