2-Bromo-3-chloro-6-fluorobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC501768-1G
PC501768-250MG
APOSPC501768-1GEA
16340
CZK
APOSPC501768-1G
APOSPC501768-250MG
2-Bromo-3-chloro-6-fluorobenzoic acid
2-Bromo-3-chloro-6-fluorobenzoic acid
Vzorec:
C₇H₃BrClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28141929 CAS číslo: 1805575-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...