Methyl 2-bromo-3-chloro-6-fluorobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC501762-1G
PC501762-250MG
APOSPC501762-1GEA
4960
CZK
APOSPC501762-1G
APOSPC501762-250MG
Methyl 2-bromo-3-chloro-6-fluorobenzoate
Methyl 2-bromo-3-chloro-6-fluorobenzoate
Vzorec:
C₈H₅BrClFO₂ MW: 267,48 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28141930 CAS číslo: 1805210-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...