2-Bromo-5-(difluoromethoxy)aniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC501748-1G
PC501748-250MG
APOSPC501748-1GEA
1660
CZK
APOSPC501748-1G
APOSPC501748-250MG
2-Bromo-5-(difluoromethoxy)aniline 95%
2-Bromo-5-(difluoromethoxy)aniline
Vzorec:
C₇H₆BrF₂NO Storage Temperature: Ambient |
MDL Number:
MFCD18390858 CAS číslo: 1261875-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...