3-Bromo-2,4-difluorobenzyl alcohol 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC501723-1G
PC501723-250MG
PC501723-5G
APOSPC501723-1GEA
14490
CZK
APOSPC501723-1G
APOSPC501723-250MG
APOSPC501723-5G
3-Bromo-2,4-difluorobenzyl alcohol 95%
3-Bromo-2,4-difluorobenzyl alcohol
Vzorec:
C₇H₅BrF₂O MW: 223,015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28737915 CAS číslo: 1499465-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...