Tisk…

3’-Bromo-2’,4’-difluoroacetophenone 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(3-Bromo-2,4-difluorophenyl)ethanone

PC501684-1G PC501684-5G PC501684-25G
APOSPC501684-1GEA 622 CZK
APOSPC501684-1G APOSPC501684-5G APOSPC501684-25G
3’-Bromo-2’,4’-difluoroacetophenone 95%
3’-Bromo-2’,4’-difluoroacetophenone
Vzorec: C₈H₅BrF₂O
MW: 235,026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13689251
CAS číslo: 1210824-63-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR