Tisk…

Methyl 3-fluoro-2-formylbenzoate

Dodavatel: Apollo Scientific
Varování

PC501662-1G PC501662-250MG
APOSPC501662-1GEA 12400 CZK
APOSPC501662-1G APOSPC501662-250MG
Methyl 3-fluoro-2-formylbenzoate
Methyl 3-fluoro-2-formylbenzoate
Vzorec: C₉H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD24685410
CAS číslo: 97711-50-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR