Methyl 4-fluoro-3-(hydroxymethyl)benzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC501655-100MG
PC501655-1G
PC501655-250MG
APOSPC501655-100MGEA
9030
CZK
APOSPC501655-100MG
APOSPC501655-1G
APOSPC501655-250MG
Methyl 4-fluoro-3-(hydroxymethyl)benzoate 95%
Methyl 4-fluoro-3-(hydroxymethyl)benzoate
Vzorec:
C₉H₉FO₃ MW: 184,164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15527250 CAS číslo: 816449-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...