2-Fluoro-3-nitrobenzenesulphonamide 95%
Dodavatel: Apollo Scientific
PC501653-1G
PC501653-250MG
APOSPC501653-1GEA
27080
CZK
APOSPC501653-1G
APOSPC501653-250MG
2-Fluoro-3-nitrobenzenesulphonamide 95%
2-Fluoro-3-nitrobenzenesulphonamide
Vzorec:
C₆H₅FN₂O₄S MW: 220,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28798466 CAS číslo: 1804044-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...