Tisk…

3-Fluoro-2-formylbenzoic acid 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC501640-1G PC501640-250MG
APOSPC501640-1GEA 15640 CZK
APOSPC501640-1G APOSPC501640-250MG
3-Fluoro-2-formylbenzoic acid 95%
3-Fluoro-2-formylbenzoic acid

Exists in equilibrium with cyclised form - 4-fluoro-3-hydroxy-2-benzofuran-1(3H)-one

Vzorec: C₈H₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD18453664
CAS číslo: 1269496-37-8
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR