5-Fluoro-2-formylbenzoic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC501602-1G
PC501602-250MG
PC501602-5G
APOSPC501602-1GEA
4730
CZK
APOSPC501602-1G
APOSPC501602-250MG
APOSPC501602-5G
5-Fluoro-2-formylbenzoic acid 99%
5-Fluoro-2-formylbenzoic acid
Exists in equilibrium with cyclised form - 6-fluoro-3-hydroxy-2-benzofuran-1(3H)-one
Vzorec:
C₈H₅FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11053106 CAS číslo: 920481-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...