Tisk…

3-(Pentafluorothio)benzeneboronic acid

Dodavatel: Apollo Scientific

PC501600-1G PC501600-250MG PC501600-5G
APOSPC501600-1GEA 20470 CZK
APOSPC501600-1G APOSPC501600-250MG APOSPC501600-5G
3-(Pentafluorothio)benzeneboronic acid
3-(Pentafluorothio)benzeneboronic acid
Vzorec: C₆H₆BF₅O₂S
MW: 247,979 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27918528
CAS číslo: 871507-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR