Tisk…

2-Bromo-4,6-difluorobenzoic acid 95%

Dodavatel: Apollo Scientific
Varování

PC501567-10G PC501567-1G PC501567-5G
APOSPC501567-10GEA 33060 CZK
APOSPC501567-10G APOSPC501567-1G APOSPC501567-5G
2-Bromo-4,6-difluorobenzoic acid 95%
2-Bromo-4,6-difluorobenzoic acid
Vzorec: C₇H₃BrF₂O₂
MW: 236,998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713104
CAS číslo: 651026-99-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR