Tisk…

3,4-Difluoro-1-(trifluoromethoxy)benzene 99%

Dodavatel: Apollo Scientific
Varování

PC501506-1G PC501506-250MG PC501506-5G
APOSPC501506-1GEA 6850 CZK
APOSPC501506-1G APOSPC501506-250MG APOSPC501506-5G
3,4-Difluoro-1-(trifluoromethoxy)benzene 99%
3,4-Difluoro-1-(trifluoromethoxy)benzene
Vzorec: C₇H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD13194832
CAS číslo: 1378671-81-8
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR