2,6-Difluoro-1-(trifluoromethoxy)benzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1,3-Difluoro-2-(trifluoromethoxy)benzene
PC501504-1G
PC501504-5G
APOSPC501504-1GEA
910
CZK
APOSPC501504-1G
APOSPC501504-5G
2,6-Difluoro-1-(trifluoromethoxy)benzene
2,6-Difluoro-1-(trifluoromethoxy)benzene
Vzorec:
C₇H₃F₅O MW: 198,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226643 CAS číslo: 153338-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...