Tisk…

2,3-Difluoro-1-(trifluoromethoxy)benzene 99%

Dodavatel: Apollo Scientific
Varování

PC501494-1G PC501494-5G PC501494-25G
APOSPC501494-1GEA 910 CZK
APOSPC501494-1G APOSPC501494-5G APOSPC501494-25G
2,3-Difluoro-1-(trifluoromethoxy)benzene 99%
2,3-Difluoro-1-(trifluoromethoxy)benzene
Vzorec: C₇H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD18641903
CAS číslo: 158178-35-9
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR