Methyl 2-fluoro-3-iodobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC501473-1G
PC501473-5G
PC501473-25G
APOSPC501473-1GEA
2030
CZK
APOSPC501473-1G
APOSPC501473-5G
APOSPC501473-25G
Methyl 2-fluoro-3-iodobenzoate
Methyl 2-fluoro-3-iodobenzoate
Vzorec:
C₈H₆FIO₂ MW: 280,035 g/mol Bod varu: 67…69 °C (2 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD12032199 CAS číslo: 1260830-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...