Tisk…

2-Bromophenylsulphur pentafluoride 98%

Dodavatel: Apollo Scientific

PC501463-1G PC501463-250MG
APOSPC501463-1GEA 13230 CZK
APOSPC501463-1G APOSPC501463-250MG
2-Bromophenylsulphur pentafluoride 98%
2-Bromophenylsulphur pentafluoride
Vzorec: C₆H₄BrF₅S
Storage Temperature: Ambient
MDL Number: MFCD26408179
CAS číslo: 1435807-16-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR