Methyl 4-(bromomethyl)-3-fluorobenzoate 95%
Dodavatel: Apollo Scientific
PC501461-1G
PC501461-5G
APOSPC501461-1GEA
2320
CZK
APOSPC501461-1G
APOSPC501461-5G
Methyl 4-(bromomethyl)-3-fluorobenzoate 95%
Methyl 4-(bromomethyl)-3-fluorobenzoate
Vzorec:
C₉H₈BrFO₂ MW: 247,061 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00273345 CAS číslo: 128577-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...