2-Bromo-3-fluoro-4-iodoaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC501459-1G
PC501459-250MG
PC501459-5G
APOSPC501459-1GEA
13460
CZK
APOSPC501459-1G
APOSPC501459-250MG
APOSPC501459-5G
2-Bromo-3-fluoro-4-iodoaniline
2-Bromo-3-fluoro-4-iodoaniline
Vzorec:
C₆H₄BrFIN MW: 315,91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28397846 CAS číslo: 1936364-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...