3-Fluoro-5-nitrobenzyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(Bromomethyl)-3-fluoro-5-nitrobenzene
PC501426-1G
PC501426-5G
APOSPC501426-1GEA
11580
CZK
APOSPC501426-1G
APOSPC501426-5G
3-Fluoro-5-nitrobenzyl bromide
3-Fluoro-5-nitrobenzyl bromide
Vzorec:
C₇H₅BrFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13185623 CAS číslo: 883987-75-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...