Tisk…

2-Amino-5-bromo-3-fluorobenzoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-2-carboxy-6-fluoroaniline

PC501399-1G PC501399-250MG PC501399-5G
APOSPC501399-1GEA 1080 CZK
APOSPC501399-1G APOSPC501399-250MG APOSPC501399-5G
2-Amino-5-bromo-3-fluorobenzoic acid
2-Amino-5-bromo-3-fluorobenzoic acid
Vzorec: C₇H₅BrFNO₂
MW: 234,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035659
CAS číslo: 874784-14-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR