5'-Fluoro-2'-(methylsulphonyl)acetophenone
Dodavatel: Apollo Scientific
|
Varování
|
PC501388-1G
PC501388-250MG
APOSPC501388-1GEA
11910
CZK
APOSPC501388-1G
APOSPC501388-250MG
5'-Fluoro-2'-(methylsulphonyl)acetophenone
5'-Fluoro-2'-(methylsulphonyl)acetophenone
Vzorec:
C₉H₉FO₃S MW: 216,229 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28348626 CAS číslo: 1845690-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...