Tisk…

3'-Fluoro-4'-(methylsulphonyl)acetophenone

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(3-Fluoro-4-methylsulphonylphenyl)ethanone

PC501386-1G PC501386-250MG
APOSPC501386-1GEA 12340 CZK
APOSPC501386-1G APOSPC501386-250MG
3'-Fluoro-4'-(methylsulphonyl)acetophenone
3'-Fluoro-4'-(methylsulphonyl)acetophenone
Vzorec: C₉H₉FO₃S
MW: 216.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441520
CAS číslo: 13372-52-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR