2-Fluoro-4-(methylsulphonyl)benzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
PC501373-1G
PC501373-5G
APOSPC501373-1GEA
2780
CZK
APOSPC501373-1G
APOSPC501373-5G
2-Fluoro-4-(methylsulphonyl)benzaldehyde
2-Fluoro-4-(methylsulphonyl)benzaldehyde
Vzorec:
C₈H₇FO₃S MW: 202,203 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13191594 CAS číslo: 1197193-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...