Tisk…

2-Fluoro-4-(methylsulphonyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

PC501371-1G PC501371-250MG
APOSPC501371-1GEA 10350 CZK
APOSPC501371-1G APOSPC501371-250MG
2-Fluoro-4-(methylsulphonyl)benzonitrile
2-Fluoro-4-(methylsulphonyl)benzonitrile
Vzorec: C₈H₆FNO₂S
MW: 199.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18378490
CAS číslo: 411233-40-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR