Tisk…

4-Bromo-3-fluorophenacylamine hydrochloride, tech

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-1-(4-bromo-3-fluorophenyl)ethanone hydrochloride

PC501322-1G PC501322-250MG
APOSPC501322-1GEA 5790 CZK
APOSPC501322-1G APOSPC501322-250MG
4-Bromo-3-fluorophenacylamine hydrochloride, tech
4-Bromo-3-fluorophenacylamine hydrochloride
Vzorec: C₈H₇BrFNO·ClH
Storage Temperature: Ambient
MDL Number: MFCD11847050
CAS číslo: 1260679-52-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR