4-Bromo-3-fluorophenacyl bromide 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC501321-1G
PC501321-250MG
PC501321-5G
APOSPC501321-1GEA
8340
CZK
APOSPC501321-1G
APOSPC501321-250MG
APOSPC501321-5G
4-Bromo-3-fluorophenacyl bromide 97%
4-Bromo-3-fluorophenacyl bromide
Vzorec:
C₈H₅Br₂FO Storage Temperature: Ambient |
MDL Number:
MFCD11847180 CAS číslo: 1003879-02-4 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...