4-Bromo-2-fluoro-6-iodoaniline
Dodavatel: Apollo Scientific
|
Varování
|
PC501279-1G
PC501279-250MG
PC501279-5G
APOSPC501279-1GEA
1660
CZK
APOSPC501279-1G
APOSPC501279-250MG
APOSPC501279-5G
4-Bromo-2-fluoro-6-iodoaniline
4-Bromo-2-fluoro-6-iodoaniline
Vzorec:
C₆H₄BrFIN MW: 315,91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22570400 CAS číslo: 1201149-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...