2-Fluoro-4-(2,2,2-trifluoroethoxy)aniline 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC501248-1G
PC501248-250MG
APOSPC501248-1GEA
24630
CZK
APOSPC501248-1G
APOSPC501248-250MG
2-Fluoro-4-(2,2,2-trifluoroethoxy)aniline 95%
2-Fluoro-4-(2,2,2-trifluoroethoxy)aniline
Vzorec:
C₈H₇F₄NO MW: 209,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16074816 CAS číslo: 1248331-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...