2-(2,2-Difluoroethoxy)aniline
Dodavatel: Apollo Scientific
|
Varování
|
PC501234-1G
PC501234-5G
APOSPC501234-1GEA
910
CZK
APOSPC501234-1G
APOSPC501234-5G
2-(2,2-Difluoroethoxy)aniline
2-(2,2-Difluoroethoxy)aniline
Vzorec:
C₈H₉F₂NO MW: 173,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08700290 CAS číslo: 937606-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...