Tisk…

6-Bromo-4-fluoro-1,3-benzoxazole

Dodavatel: Apollo Scientific
Varování

PC501231-1G PC501231-250MG
APOSPC501231-1GEA 11180 CZK
APOSPC501231-1G APOSPC501231-250MG
6-Bromo-4-fluoro-1,3-benzoxazole
6-Bromo-4-fluoro-1,3-benzoxazole
Vzorec: C₇H₃BrFNO
MW: 216,007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28348528
CAS číslo: 1935181-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR