2-Amino-5-bromo-3-fluorophenol
Dodavatel: Apollo Scientific
|
Varování
|
PC501229-1G
PC501229-5G
PC501229-10G
APOSPC501229-1GEA
2400
CZK
APOSPC501229-1G
APOSPC501229-5G
APOSPC501229-10G
2-Amino-5-bromo-3-fluorophenol
2-Amino-5-bromo-3-fluorophenol
Vzorec:
C₆H₅BrFNO Storage Temperature: Ambient |
MDL Number:
MFCD28348526 CAS číslo: 186309-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...