Tisk…

3'-Fluoro-2'-nitroacetophenone 98%

Dodavatel: Apollo Scientific
Varování

PC501159-1G PC501159-5G
APOSPC501159-1GEA 4070 CZK
APOSPC501159-1G APOSPC501159-5G
3'-Fluoro-2'-nitroacetophenone 98%
3'-Fluoro-2'-nitroacetophenone
Vzorec: C₈H₆FNO₃
Storage Temperature: Ambient
MDL Number: MFCD13194364
CAS číslo: 1214346-37-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR