3-Fluoro-2-nitrobenzyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC501144-1G
PC501144-250MG
PC501144-5G
APOSPC501144-1GEA
13460
CZK
APOSPC501144-1G
APOSPC501144-250MG
APOSPC501144-5G
3-Fluoro-2-nitrobenzyl bromide 95%
3-Fluoro-2-nitrobenzyl bromide
Vzorec:
C₇H₅BrFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13185622 CAS číslo: 1104543-73-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...