Tisk…

4-Amino-2-fluoro-3-methylbenzonitrile

Dodavatel: Apollo Scientific
Varování

PC501107-1G PC501107-250MG PC501107-5G
APOSPC501107-1GEA 1660 CZK
APOSPC501107-1G APOSPC501107-250MG APOSPC501107-5G
4-Amino-2-fluoro-3-methylbenzonitrile
4-Amino-2-fluoro-3-methylbenzonitrile
Vzorec: C₈H₇FN₂
MW: 150,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18826339
CAS číslo: 757247-93-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR