3-Bromo-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-4-[methoxy(methyl)carbamoyl]benzotrifluoride
PC50107-1G
PC50107-250MG
PC50107-5G
APOSPC50107-1GEA
15540
CZK
APOSPC50107-1G
APOSPC50107-250MG
APOSPC50107-5G
3-Bromo-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
3-Bromo-N-methoxy-N-methyl-4-(trifluoromethyl)benzamide
Vzorec:
C₁₀H₉BrF₃NO₂ MW: 312,083 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28515423 CAS číslo: 1820664-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...