Tisk…

4-Bromo-3-fluoro-6-nitro-2-(trifluoromethyl)aniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-5-bromo-6-fluoro-3-nitrobenzotrifluoride

PC50106-1G PC50106-250MG PC50106-5G
APOSPC50106-1GEA 10750 CZK
APOSPC50106-1G APOSPC50106-250MG APOSPC50106-5G
4-Bromo-3-fluoro-6-nitro-2-(trifluoromethyl)aniline
4-Bromo-3-fluoro-6-nitro-2-(trifluoromethyl)aniline
Vzorec: C₇H₃BrF₄N₂O₂
MW: 303,009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28515422
CAS číslo: 1820687-67-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR